Tisk…

N-tert-Butyl-2-methylenesuccinamic acid

Dodavatel: Apollo Scientific

OR346120-1G OR346120-5G
APOSOR346120-1GEA 0 CZK
APOSOR346120-1G APOSOR346120-5G
N-tert-Butyl-2-methylenesuccinamic acid
N-tert-Butyl-2-methylenesuccinamic acid
Vzorec: C₉H₁₅NO₃
MW: 185,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11227162
CAS číslo: 1089330-58-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR