Tisk…
Methyl-6-methyl-1H-indole-2-carboxylate
Dodavatel: Apollo Scientific
Synonyms: 6-Methyl-1H-indole-2-carboxylic acid methyl ester, Methyl 6-methyl-1H-indole-2-carboxylate
OR346107-1G OR346107-500MG OR346107-5G
APOSOR346107-1GEA 0 CZK
APOSOR346107-1G APOSOR346107-500MG APOSOR346107-5G
Methyl-6-methyl-1H-indole-2-carboxylate
Methyl-6-methyl-1H-indole-2-carboxylate
Vzorec: C₁₁H₁₁NO₂
MW: 189,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06653197
CAS číslo: 18377-65-6