Tisk…

(4-Methylpiperazin-1-yl)phenylacetonitrile hydrochloride

Dodavatel: Apollo Scientific

OR346105-1G OR346105-5G
APOSOR346105-1GEA 0 CZK
APOSOR346105-1G APOSOR346105-5G
(4-Methylpiperazin-1-yl)phenylacetonitrile hydrochloride
(4-Methylpiperazin-1-yl)phenylacetonitrile hydrochloride
Vzorec: C₁₃H₁₈ClN₃
MW: 251,76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052463
CAS číslo: 108953-41-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR