Tisk…

4-Hydroxymethyl-2-pyridin-2-yl-3H-imidazole

Dodavatel: Apollo Scientific

OR346082-1G OR346082-5G
APOSOR346082-1GEA 0 CZK
APOSOR346082-1G APOSOR346082-5G
4-Hydroxymethyl-2-pyridin-2-yl-3H-imidazole
4-Hydroxymethyl-2-pyridin-2-yl-3H-imidazole
Vzorec: C₉H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD10568331
CAS číslo: 53292-71-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR