Tisk…

5-Aminomethyl-3-pyridin-3-yl[1,2,4]oxadiazole dihydrochloride

Dodavatel: Apollo Scientific

OR346075-1G OR346075-5G
APOSOR346075-1GEA 0 CZK
APOSOR346075-1G APOSOR346075-5G
5-Aminomethyl-3-pyridin-3-yl[1,2,4]oxadiazole dihydrochloride
5-Aminomethyl-3-pyridin-3-yl[1,2,4]oxadiazole dihydrochloride
Vzorec: C₈H₁₀Cl₂N₄O
MW: 249,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15142769
CAS číslo: 1208081-52-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR