2-(3-Methoxyphenyl)thioacetamide
Dodavatel: Apollo Scientific
OR346069-1G
OR346069-5G
APOSOR346069-1GEA
0
CZK
APOSOR346069-1G
APOSOR346069-5G
2-(3-Methoxyphenyl)thioacetamide
2-(3-Methoxyphenyl)thioacetamide
Vzorec:
C₉H₁₁NOS MW: 181,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09414738 CAS číslo: 35582-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...