Tisk…
2-Methoxybenzamidine hydrochloride
Dodavatel: Apollo Scientific
OR346059-1G OR346059-5G
APOSOR346059-1GEA 0 CZK
APOSOR346059-1G APOSOR346059-5G
2-Methoxybenzamidine hydrochloride
2-Methoxybenzamidine hydrochloride
Vzorec: C₈H₁₁ClN₂O
MW: 186.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738644
CAS číslo: 51818-19-0