Tisk…

N'-[1-Aminopropylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346054-1G OR346054-500MG
APOSOR346054-1GEA 0 CZK
APOSOR346054-1G APOSOR346054-500MG
N'-[1-Aminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Aminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₈H₁₇N₃O₂
MW: 187,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568290
CAS číslo: 159016-21-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR