N'-[1-Cyclobutylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
OR346044-1G
OR346044-500MG
APOSOR346044-1GEA
0
CZK
APOSOR346044-1G
APOSOR346044-500MG
N'-[1-Cyclobutylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclobutylaminopropylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₂H₂₃N₃O₂ MW: 241,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568258 CAS číslo: 1053657-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...