Tisk…

N'-[1-Cyclopentylamino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346041-1G OR346041-500MG
APOSOR346041-1GEA 0 CZK
APOSOR346041-1G APOSOR346041-500MG
N'-[1-Cyclopentylamino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclopentylamino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₉H₂₉N₃O₃
MW: 347,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568254
CAS číslo: 1053657-57-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR