Tisk…

N'-[1-Amino-1-phenylmethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346040-1G OR346040-500MG
APOSOR346040-1GEA 0 CZK
APOSOR346040-1G APOSOR346040-500MG
N'-[1-Amino-1-phenylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-phenylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₂H₁₇N₃O₂
MW: 235,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568253
CAS číslo: 159016-25-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR