Tisk…

N'-[2-(4-Chlorophenyl)-1-cyclobutylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346038-1G OR346038-500MG
APOSOR346038-1GEA 0 CZK
APOSOR346038-1G APOSOR346038-500MG
N'-[2-(4-Chlorophenyl)-1-cyclobutylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[2-(4-Chlorophenyl)-1-cyclobutylaminoethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₇H₂₄ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD10568251
CAS číslo: 1053657-48-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR