N'-[1-Cyclopropylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
OR346035-1G
OR346035-500MG
APOSOR346035-1GEA
0
CZK
APOSOR346035-1G
APOSOR346035-500MG
N'-[1-Cyclopropylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Cyclopropylamino-2-phenylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₆H₂₃N₃O₂ MW: 289,38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568248 CAS číslo: 1053657-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...