Tisk…

N'-[1-Amino-1-(4-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346017-1G OR346017-500MG
APOSOR346017-1GEA 0 CZK
APOSOR346017-1G APOSOR346017-500MG
N'-[1-Amino-1-(4-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-(4-chlorophenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₂H₁₆ClN₃O₂
MW: 269,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568197
CAS číslo: 1053656-11-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR