Tisk…

N'-[1-Amino-2-(3,5-dichlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346016-1G OR346016-500MG
APOSOR346016-1GEA 0 CZK
APOSOR346016-1G APOSOR346016-500MG
N'-[1-Amino-2-(3,5-dichlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(3,5-dichlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₃H₁₇Cl₂N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD10568196
CAS číslo: 1053656-08-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR