Tisk…

N'-[1-Amino-2-(2,6-dichlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346014-1G OR346014-500MG
APOSOR346014-1GEA 0 CZK
APOSOR346014-1G APOSOR346014-500MG
N'-[1-Amino-2-(2,6-dichlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(2,6-dichlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₃H₁₇Cl₂N₃O₂
MW: 318,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568194
CAS číslo: 1053656-02-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR