Tisk…

N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346008-1G OR346008-500MG
APOSOR346008-1GEA 0 CZK
APOSOR346008-1G APOSOR346008-500MG
N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(4-methoxyphenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₄H₂₁N₃O₃
MW: 279,34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568184
CAS číslo: 1053655-79-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR