Tisk…

N'-[1-Amino-1-p-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester

Dodavatel: Apollo Scientific

OR346006-1G OR346006-500MG
APOSOR346006-1GEA 0 CZK
APOSOR346006-1G APOSOR346006-500MG
N'-[1-Amino-1-p-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-p-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec: C₁₃H₁₉N₃O₂
MW: 249,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568182
CAS číslo: 1053655-75-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR