N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
OR346004-1G
OR346004-500MG
APOSOR346004-1GEA
0
CZK
APOSOR346004-1G
APOSOR346004-500MG
N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-(4-chlorophenyl)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₃H₁₈ClN₃O₂ MW: 283,76 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568180 CAS číslo: 159016-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...