N'-[1-Amino-1-m-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
OR346003-1G
OR346003-500MG
APOSOR346003-1GEA
0
CZK
APOSOR346003-1G
APOSOR346003-500MG
N'-[1-Amino-1-m-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-1-m-tolylmethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₃H₁₉N₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD10568178 CAS číslo: 1053655-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...