N'-[1-Amino-2-p-tolylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
OR346002-1G
OR346002-500MG
APOSOR346002-1GEA
0
CZK
APOSOR346002-1G
APOSOR346002-500MG
N'-[1-Amino-2-p-tolylethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-p-tolylethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₄H₂₁N₃O₂ MW: 263,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568177 CAS číslo: 159016-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...