N'-[1-Amino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
Dodavatel: Apollo Scientific
OR346000-1G
OR346000-500MG
APOSOR346000-1GEA
0
CZK
APOSOR346000-1G
APOSOR346000-500MG
N'-[1-Amino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
N'-[1-Amino-2-phenoxyethylidene]hydrazinecarboxylic acid-tert-butyl ester
Vzorec:
C₁₃H₁₉N₃O₃ MW: 265,31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10568175 CAS číslo: 1053655-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...