N-Boc-1,6-diaminohexane 97%
Dodavatel: Apollo Scientific
OR345618-25G
OR345618-100G
APOSOR345618-25GEA
2320
CZK
APOSOR345618-25G
APOSOR345618-100G
N-Boc-1,6-diaminohexane 97%
N-Boc-1,6-diaminohexane
Vzorec:
C₁₁H₂₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00671489 CAS číslo: 51857-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...