4-Amino-5-bromo-3-methylpyridine 95+%
Dodavatel: Apollo Scientific
OR345470-25G
OR345470-5G
APOSOR345470-25GEA
9100
CZK
APOSOR345470-25G
APOSOR345470-5G
4-Amino-5-bromo-3-methylpyridine 95+%
4-Amino-5-bromo-3-methylpyridine
Vzorec:
C₆H₇BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD07437841 CAS číslo: 97944-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...