Tisk…

5-Bromo-2-chloro-3-iodopyridine 99%

Dodavatel: Apollo Scientific
Nebezpečí

OR345455-25G OR345455-5G OR345455-1G
APOSOR345455-25GEA 6420 CZK
APOSOR345455-25G APOSOR345455-5G APOSOR345455-1G
5-Bromo-2-chloro-3-iodopyridine 99%
5-Bromo-2-chloro-3-iodopyridine
Vzorec: C₅H₂BrClIN
Storage Temperature: Ambient
MDL Number: MFCD08688585
CAS číslo: 928653-73-0
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR