5-Bromo-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine 98%
Dodavatel: Apollo Scientific
OR345430-1G
OR345430-5G
APOSOR345430-1GEA
2650
CZK
APOSOR345430-1G
APOSOR345430-5G
5-Bromo-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-1-triisopropylsilanyl-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₁₆H₂₅BrN₂Si Storage Temperature: Ambient |
MDL Number:
MFCD08688597 CAS číslo: 858116-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...