2-Amino-5-bromonicotinonitrile 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Amino-5-bromo-nicotinonitrile, 2-Amino-5-bromo-3-cyanopyridine
OR345399-25G
OR345399-5G
APOSOR345399-25GEA
5360
CZK
APOSOR345399-25G
APOSOR345399-5G
2-Amino-5-bromonicotinonitrile 97%
2-Amino-5-bromonicotinonitrile
Vzorec:
C₆H₄BrN₃ MW: 198,0201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08688591 CAS číslo: 709652-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...