Tisk…

3-Nitropicolinic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Nitropyridine-2-carboxylic acid

OR345379-1G OR345379-5G
APOSOR345379-1GEA 2980 CZK
APOSOR345379-1G APOSOR345379-5G
3-Nitropicolinic acid
3-Nitropicolinic acid
Vzorec: C₆H₄N₂O₄
MW: 168,1070 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658500
CAS číslo: 59290-85-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR