3-Hydrazino-2-nitropyridine
Dodavatel: Apollo Scientific
OR345374-1G
OR345374-250MG
OR345374-500MG
OR345374-2.5G
APOSOR345374-1GEA
9330
CZK
APOSOR345374-1G
APOSOR345374-250MG
APOSOR345374-500MG
APOSOR345374-2.5G
3-Hydrazino-2-nitropyridine
3-Hydrazino-2-nitropyridine
Vzorec:
C₅H₆N₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00493934 CAS číslo: 57115-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...