2-Amino-6-chloro-5-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloro-5-methylpyridin-2-amine
OR345347-1G
OR345347-25G
OR345347-5G
APOSOR345347-1GEA
1990
CZK
APOSOR345347-1G
APOSOR345347-25G
APOSOR345347-5G
2-Amino-6-chloro-5-methylpyridine
2-Amino-6-chloro-5-methylpyridine
Vzorec:
C₆H₇ClN₂ MW: 142,5900 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13193513 CAS číslo: 442129-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...