Tisk…

2-Bromo-p-toluonitrile 98%

Dodavatel: Apollo Scientific

OR345343-100G
APOSOR345343-100GEA 3440 CZK
APOSOR345343-100G
2-Bromo-p-toluonitrile 98%
2-Bromo-p-toluonitrile
Vzorec: C₈H₆BrN
MW: 196,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00158968
CAS číslo: 42872-73-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR