3-Bromo-2-methyl-5-nitropyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR345288-10G
OR345288-5G
APOSOR345288-10GEA
3740
CZK
APOSOR345288-10G
APOSOR345288-5G
3-Bromo-2-methyl-5-nitropyridine 98%
3-Bromo-2-methyl-5-nitropyridine
Vzorec:
C₆H₅BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08688606 CAS číslo: 186593-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...