5-Nitropyridin-3-ol 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR345287-1G
OR345287-250MG
APOSOR345287-1GEA
6390
CZK
APOSOR345287-1G
APOSOR345287-250MG
5-Nitropyridin-3-ol 95%
5-Nitropyridin-3-ol
Vzorec:
C5H4N2O3 Storage Temperature: Ambient |
MDL Number:
MFCD04114224 CAS číslo: 186593-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...