Tisk…

2-Cyanoisonicotinic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR345273-1G OR345273-5G
APOSOR345273-1GEA 695 CZK
APOSOR345273-1G APOSOR345273-5G
2-Cyanoisonicotinic acid
2-Cyanoisonicotinic acid
Vzorec: C₇H₄N₂O₂
Bod tání: 236 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411729
CAS číslo: 161233-97-2
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR