Tisk…

5-Bromo-6-chloropyridin-3-ol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-2-chloro-5-hydroxypyridine

OR345245-5G
APOSOR345245-5GEA 4040 CZK
APOSOR345245-5G
5-Bromo-6-chloropyridin-3-ol 97%
5-Bromo-6-chloropyridin-3-ol
Vzorec: C₅H₃BrClNO
MW: 208,45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835248
CAS číslo: 130115-85-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR