Tisk…

2-Methoxy-6-methyl-3-nitropyridine

Dodavatel: Apollo Scientific
Varování

OR345207-1G OR345207-5G
APOSOR345207-1GEA 3640 CZK
APOSOR345207-1G APOSOR345207-5G
2-Methoxy-6-methyl-3-nitropyridine
2-Methoxy-6-methyl-3-nitropyridine
Vzorec: C₇H₈N₂O₃
MW: 168,150 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095074
CAS číslo: 112163-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR