Tisk…

5-Nitropyridine-2-carbonitrile 95+%

Dodavatel: Apollo Scientific

Synonyms: 2-Cyano-5-nitropyridine, 5-Nitropicolinonitrile

OR345166-5G OR345166-1G OR345166-25G
APOSOR345166-5GEA 7840 CZK
APOSOR345166-5G APOSOR345166-1G APOSOR345166-25G
5-Nitropyridine-2-carbonitrile 95+%
5-Nitropyridine-2-carbonitrile
Vzorec: C₆H₃N₃O₂
MW: 149,1069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368184
CAS číslo: 100367-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR