Ketorolac 95%
Dodavatel: Apollo Scientific
Synonyms:
5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid, Ketorolaco, Ketorolacum
OR345134-10G
OR345134-1G
OR345134-5G
APOSOR345134-10GEA
16530
CZK
APOSOR345134-10G
APOSOR345134-1G
APOSOR345134-5G
Ketorolac 95%
Ketorolac
Vzorec:
C₁₅H₁₃NO₃ MW: 255,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00864281 CAS číslo: 74103-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...