Tisk…

3-Hydroxy-2-nitrobenzoic acid

Dodavatel: Apollo Scientific

OR345117-10G OR345117-5G
APOSOR345117-10GEA 6550 CZK
APOSOR345117-10G APOSOR345117-5G
3-Hydroxy-2-nitrobenzoic acid
3-Hydroxy-2-nitrobenzoic acid
Vzorec: C₇H₅NO₅
Storage Temperature: Ambient
MDL Number: MFCD00017003
CAS číslo: 602-00-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR