2-Chloro-4-hydroxybenzoic acid
Dodavatel: Apollo Scientific
OR345112-25G
OR345112-5G
APOSOR345112-25GEA
9430
CZK
APOSOR345112-25G
APOSOR345112-5G
2-Chloro-4-hydroxybenzoic acid
2-Chloro-4-hydroxybenzoic acid
Vzorec:
C₇H₅ClO₃ MW: 172,57 g/mol Bod tání: 207…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045855 CAS číslo: 56363-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...