3-Acetyl-5-bromopyridine 97+%
Dodavatel: Apollo Scientific
OR345090-10G
OR345090-5G
APOSOR345090-10GEA
910
CZK
APOSOR345090-10G
APOSOR345090-5G
3-Acetyl-5-bromopyridine 97+%
3-Acetyl-5-bromopyridine
Vzorec:
C₇H₆BrNO Bod tání: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD03086033 CAS číslo: 38940-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...