3-Chloro-4-methoxybenzoic acid 98%
Dodavatel: Apollo Scientific
OR345088-25G
OR345088-1G
OR345088-5G
APOSOR345088-25GEA
2690
CZK
APOSOR345088-25G
APOSOR345088-1G
APOSOR345088-5G
3-Chloro-4-methoxybenzoic acid 98%
3-Chloro-4-methoxybenzoic acid
Vzorec:
C₈H₇ClO₃ Bod tání: 213…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016512 CAS číslo: 37908-96-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...