4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 96%
Dodavatel: Apollo Scientific
OR345081-1G
OR345081-10G
OR345081-5G
APOSOR345081-1GEA
2280
CZK
APOSOR345081-1G
APOSOR345081-10G
APOSOR345081-5G
4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 96%
4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine
Vzorec:
C₁₄H₁₁BrN₂O₂S Storage Temperature: Ambient |
MDL Number:
MFCD10575009 CAS číslo: 348640-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...