Tisk…

5-Amino-2-iodopyridine 98%

Dodavatel: Apollo Scientific

OR345071-1G OR345071-5G
APOSOR345071-1GEA 2440 CZK
APOSOR345071-1G APOSOR345071-5G
5-Amino-2-iodopyridine 98%
5-Amino-2-iodopyridine
Vzorec: C₅H₅IN₂
Storage Temperature: Ambient
MDL Number: MFCD02706663
CAS číslo: 29958-12-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR