Tisk…

7,8-Dihydroxy-4-methylcoumarin

Dodavatel: Apollo Scientific

OR345049-1G OR345049-5G
APOSOR345049-1GEA 0 CZK
APOSOR345049-1G APOSOR345049-5G
7,8-Dihydroxy-4-methylcoumarin
7,8-Dihydroxy-4-methylcoumarin
Vzorec: C₁₀H₈O₄
Storage Temperature: Ambient
MDL Number: MFCD00016972
CAS číslo: 2107-77-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR