3-Amino-2,4-dichloropyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR345038-25G
APOSOR345038-25GEA
2570
CZK
APOSOR345038-25G
3-Amino-2,4-dichloropyridine
3-Amino-2,4-dichloropyridine
Vzorec:
C5H4Cl2N2 MW: 163,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09750301 CAS číslo: 173772-63-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...