2,5-Dibromo-3-chloropyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Chloro-2,5-dibromopyridine
OR345031-1G
OR345031-5G
APOSOR345031-1GEA
622
CZK
APOSOR345031-1G
APOSOR345031-5G
2,5-Dibromo-3-chloropyridine
2,5-Dibromo-3-chloropyridine
Vzorec:
C₅H₂Br₂ClN MW: 271,3371 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08277306 CAS číslo: 160599-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...