Tisk…

3-Iodo-2-methoxypyridine 98%

Dodavatel: Apollo Scientific

OR345009-100G OR345009-25G
APOSOR345009-100GEA 7870 CZK
APOSOR345009-100G APOSOR345009-25G
3-Iodo-2-methoxypyridine 98%
3-Iodo-2-methoxypyridine
Vzorec: C₆H₆INO
MW: 235.02
Storage Temperature: Ambient
MDL Number: MFCD03094946
CAS číslo: 112197-15-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR