1-Boc-4-chloroindole-2-boronic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Chloro-N-(Boc)-indole-2-boronic acid, (1-(tert-Butoxycarbonyl)-4-chloro-1H-indol-2-yl)boronic acid
OR3437-1G
OR3437-5G
APOSOR3437-1GEA
827
CZK
APOSOR3437-1G
APOSOR3437-5G
1-Boc-4-chloroindole-2-boronic acid 98%
1-Boc-4-chloroindole-2-boronic acid
Vzorec:
C₁₃H₁₅BClNO₄ MW: 295,53 g/mol Bod tání: 107…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114582 CAS číslo: 475102-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...