7-Methyl-2-(3-nitrophenyl)imidazo[1,2-a]pyridine
Dodavatel: Apollo Scientific
OR3412-1G
OR3412-5G
APOSOR3412-1GEA
3310
CZK
APOSOR3412-1G
APOSOR3412-5G
7-Methyl-2-(3-nitrophenyl)imidazo[1,2-a]pyridine
7-Methyl-2-(3-nitrophenyl)imidazo[1,2-a]pyridine
Vzorec:
C₁₄H₁₁N₃O₂ MW: 253.26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00438465 CAS číslo: 54970-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...